Print…
Capillary columns, Rapid-MS
Fornecedor: VARIAN
CP8133 CP8121 CP8131 CP8132
VARICP8133EA 1530 EUR
VARICP8133 VARICP8121 VARICP8131 VARICP8132
Capillary columns, Rapid-MS
Cromatografia, Colunas
Rapid-MS columns reduce analysis duration by 3 to 5 times for temperature programmed, and up to 10 times for isothermal runs.
  • Fast analysis improves productivity
  • High loadability for better detection limits
  • Easy installation enhances efficiency
Rapid-MS columns utilise the high optimal carrier gas velocity obtained when a separation is performed under reduced pressure to reduce analysis times. The low bleed VF-5ms stationary phase is equivalent to a 5% phenyl, 95% dimethylpolysiloxane phase.