Print…
Masterflex® Ismatec® Footswitch for Ecoline Drives, 8-Pin DIN Connector; 6-ft (1.8 m)
Fornecedor: Avantor Fluid Handling
VWR Item#MFLX78016-95
Your Organization's Price:
Simplify your dispensing.
  • Select the footswitch with connectio suitable for your Ismatec® pump
Footswitches provide convenient, hands-free operation of your pump drives from up to six feet away. Simply press once to start pump, press again to stop.