Masterflex® PTFE Multi-Turn Needle Valve, 1/4" OD, Al Body, 2800 mL/min water
Fornecedor: Avantor Fluid Handling
Easily meter corrosive fluids or gases.
- Constructed of durable PTFE
Maximum pressure is 75 psi; maximum temperature is +200 °F (+93 °C). Wetted parts are body and O-ring made of PTFE and CTFE valve spindle. Choose aluminum (Al) or 316 stainless steel (SS) body. Orifice is 0.125 inch.