Print…
Thermo Scientific Nalgene 489 Linear LDPE Tubing, 5/8" ID x 3/4" OD; 100 Ft
Fornecedor: Avantor Fluid Handling
VWR Item#MFLX95912-06
Your Organization's Price:
Suitable for food processing applications.
  • Global market leader in range of formulations for most extensive chemical compatibility
Thermo Scientific Nalgene 489 linear low-density polyethylene (LLDPE) is a general purpose tubing suitable for many applications. Made of FDA compliant resin, is it suitable for food processing applications as well as general laboratory or industrial use. 489 LLDPE offers broad chemical resistance and is more flexible than other polyethylene formulations. This translucent formulation is nontoxic, odorless, and tasteless. The tubing may be sterilized by ethylene oxide. It is usable from -100 to 176 °F (-73 to 80 °C).