Explosion-Proof Single-Phase TEFC Motor, 56C, 0.25 HP/1800 rpm; 115/208-230 V
Fornecedor: Avantor Fluid Handling
For Class 1 Group C and D, Class 2 Group F and G locations.
- Automatic overload protection stops and restarts motor in case of overload or overheating
- UL-listed file no. E-12044
Motors are TEFC (models 70007-00 to -12 are TENV) and include an explosion-proof conduit box (shipped loose).