Print…
McMillan Flow High-Accuracy Flowmeter with Display, Stainless Steel, 0 to 5 VDC, 0.5 to 5 LPM
Fornecedor: Avantor Fluid Handling
VWR Item#MFLX32718-30
Your Organization's Price:
Economical design with an outstanding ±1.0 % full-scale accuracy.
  • A wide range of outputs options for integration into any system
Select these flowmeters and transmitters for extremely low flow-rate applications or for higher flow rate measurements and yet maintain a full-scale accuracy of ±1% or better. All units feature a stainless steel housing and come standard with stainless steel compression fittings.<br /><br />The flowmeters/transmitters can be used with a wide variety of transparent, low-viscosity (under 10 cSt) corrosive or neutral liquids and solvents. <br /><br /><B>Note:</B> A power supply is <B>required</B> for each meter/ transmitter and may be ordered separately. If using an existing power supply, order cable assembly 32704-52.
Choose transmitters with a 0 to 5 VDC or 4 to 20 mA output for control or datalogging purposes. Models 32718-20 to -32 offer a single output (0 to 5 VDC) along with a built-in 31/2-digit display.