Print…
Power selector, WRT00116
Fornecedor: WINKLER
WRT00116-XX0CH WRT00116-XX013 WRT00116-XX0UK
WINKWRT00116-XX0CHEA 113 EUR
WINKWRT00116-XX0CH WINKWRT00116-XX013 WINKWRT00116-XX0UK
Power selector, WRT00116
Controladores Controladores de temperatura
Plug-in power selector made of PS with switching mechanism.
  • Rotary switch with bimetal element
  • Signal light for controlling status indication
  • Cable with grounding plug type