Print…
Whatman™ Mini-UniPrep™ Multi-Compressor, Whatman products (Cytiva)
Fornecedor: Whatman products (Cytiva)
MUPMCPBC8
WHATMUPMCPBC8EA 366 EUR
WHATMUPMCPBC8
Whatman™ Mini-UniPrep™ Multi-Compressor, Whatman products (Cytiva)
Prensas
Mini-UniPrep™ multicompressor including one 8-position tray for standard plastic Mini-UniPrep™ filter vials.
  • Streamlines syringeless filtration by filtering up to eight filter vials simultaneously
  • Designed for Cytiva's Mini-UniPrep™ standard plastic filter vial
  • Mini-UniPrep™ vials can be loaded directly into the autosampler
The newly redesigned Whatman™ Mini-UniPrep™ multicompressor streamlines filtration, saves time and reduces hand stress by enabling the filtration of eight Mini-UniPrep™ vials at once.
Informações de encomenda: Additional traysMais informação do produto