Print…
Cartão de colheita de amostras Whatman™ 903 Proteinsaver
Fornecedor: Whatman products (Cytiva)
10531018
WHAT10531018EA 191 EUR
WHAT10531018
Cartão de colheita de amostras Whatman™ 903 Proteinsaver
Sistemas para diagnóstico Clinical Sample Collection/Preparation Coleta/Preparação de Amostras Biológicas
Whatman 903 sample collection cards are designed for the collection of biological fluids.
  • Ready to use cards saving time during sample collection, transport, and archiving
  • Strict quality control and ISO 9001 manufacturing standards ensure high quality, reproducibility, and purity
  • Choice of five-spot Proteinsaver card or four-spot Proteinsaver snap-apart card
The sample collection area contains five circles, each circle holds 75 to 80 µl of sample. Wraparound cover has spaces for name and date of collection and is imprinted with the biohazard symbol.Mais informação do produto