Print…
Toximaster® QC8 software
Fornecedor: FUJIFILM WAKO CHEMICALS
299-36161 297-36221 295-36141 294-36231
WAKF299-36161EA 9920 EUR
WAKF299-36161 WAKF297-36221 WAKF295-36141 WAKF294-36231
Toximaster® QC8 software
Software Programas de Laboratório
The Toximaster® QC8 software supports data processing based on the protocols complying with three types of pharmacopeia (USP, EP, JP) for bacterial endotoxin testing. This software is compliant with FDA 21 CFR Part 11 ERES (electronic records/electronic signature), which requires the proper information from all parties involved in the testing of the sample.
  • Endotoxin determination in compliance with pharmacopoeias (USP/EP/JP) for BET
  • Capable of creating three types of curves: internal standard curve, manual input curve and a measured curve
  • Product
Mais informação do produto