Print…
Uniform latex particles for immobilisation of biomolecules, PL-Latex
Fornecedor: VARIAN
PL6000-7101 PL6000-7102 PL6000-7103 PL6001-4101 PL6001-4102 PL6001-4103 PL6002-2101 PL6002-2102 PL6002-2103 PL6003-2101 PL6003-2102 PL6003-2103 PL6003-2121 PL6003-2122 PL6003-2123 PL6004-4101 PL6004-4102 PL6004-4103 PL6006-4101 PL6006-4102 PL6006-4103 PL6006-4161 PL6006-4162 PL6006-4163 PL6008-4101 PL6008-4102 PL6008-4103 PL6008-4121 PL6008-4122 PL6008-4123 PL6008-4141 PL6008-4143 PL6010-4101 PL6010-4102 PL6010-4103 PL6104-6101 PL6104-6121 PL6104-6122 PL6104-6141 PL6104-6142 PL6115-6101 PL6115-6102 PL6115-6103 PL6202-6101 PL6202-6102 PL6202-6103 PL6202-6121 PL6202-6122 PL6203-6101 PL6203-6102 PL6203-6103 PL6203-6121 PL6203-6122 PL6203-6123 PL6203-6131 PL6203-6132 PL6203-6141 PL6203-6143 PL6203-6181 PL6203-6182 PL6204-6101 PL6204-6102 PL6204-6103 PL6204-6141 PL6208-6101 PL6208-6102 PL6208-6103 PL6212-5101 PL6212-5102 PL6212-5103 PL6215-6101 PL6215-6102 PL6215-6103
VARIPL6000-7101EA 390 EUR
VARIPL6000-7101 VARIPL6000-7102 VARIPL6000-7103 VARIPL6001-4101 VARIPL6001-4102 VARIPL6001-4103 VARIPL6002-2101 VARIPL6002-2102 VARIPL6002-2103 VARIPL6003-2101 VARIPL6003-2102 VARIPL6003-2103 VARIPL6003-2121 VARIPL6003-2122 VARIPL6003-2123 VARIPL6004-4101 VARIPL6004-4102 VARIPL6004-4103 VARIPL6006-4101 VARIPL6006-4102 VARIPL6006-4103 VARIPL6006-4161 VARIPL6006-4162 VARIPL6006-4163 VARIPL6008-4101 VARIPL6008-4102 VARIPL6008-4103 VARIPL6008-4121 VARIPL6008-4122 VARIPL6008-4123 VARIPL6008-4141 VARIPL6008-4143 VARIPL6010-4101 VARIPL6010-4102 VARIPL6010-4103 VARIPL6104-6101 VARIPL6104-6121 VARIPL6104-6122 VARIPL6104-6141 VARIPL6104-6142 VARIPL6115-6101 VARIPL6115-6102 VARIPL6115-6103 VARIPL6202-6101 VARIPL6202-6102 VARIPL6202-6103 VARIPL6202-6121 VARIPL6202-6122 VARIPL6203-6101 VARIPL6203-6102 VARIPL6203-6103 VARIPL6203-6121 VARIPL6203-6122 VARIPL6203-6123 VARIPL6203-6131 VARIPL6203-6132 VARIPL6203-6141 VARIPL6203-6143 VARIPL6203-6181 VARIPL6203-6182 VARIPL6204-6101 VARIPL6204-6102 VARIPL6204-6103 VARIPL6204-6141 VARIPL6208-6101 VARIPL6208-6102 VARIPL6208-6103 VARIPL6212-5101 VARIPL6212-5102 VARIPL6212-5103 VARIPL6215-6101 VARIPL6215-6102 VARIPL6215-6103
Uniform latex particles for immobilisation of biomolecules, PL-Latex
Extracção de proteínas Reagentes de Extração de Proteínas
Uniform latex particles are used in a wide range of applications that exploit their physico-chemical properties in sophisticated immunoassay formats, both in medicine and veterinary science research.
Manufacture latexes with essentially uncharged surfaces for immobilisation of biomolecules by adsorption, and surfaces having carboxyl groups, allowing covalent immobilisation of ligands.
Latex particles have been employed for many years in a variety of assay formats for rapid, user-friendly and cost-effective tests, including slide agglutination, membrane migration, particle capture, turbidimetry, nephelometry, and “particle enhanced” assays.
The intensely coloured HiDye™ particles offer all the features of the white particles with the added advantage of intense colour. As the dye is not surface bound, it does not interfere with the binding of the ligand. HiDye™ particles are ideal for membrane migration assays and cell labelling studies.