Print…
UV mini lamps
Fornecedor: UVP ULTRA VIOLET PRODUCTS
95-0158-04 95-0125-05 95-0305-03 95-0188-02
UVPA95-0158-04EA 160 EUR
UVPA95-0158-04 UVPA95-0125-05 UVPA95-0305-03 UVPA95-0188-02
UV mini lamps
Luzes UV, Lâmpadas
These lamps are excellent for quick field inspection or fluorescence applications.
  • For field use and fluorescence work
  • Longwave, shortwave, and combination options available
  • Four-watt intensity
  • Impact-resistant plastic
Mini UV Lamps feature four-watt intensity and are powered by four AA batteries (not included). Each lamp housing is made of impact-resistant plastic that resists fading or cracking. The lamps are lightweight and small enough to fit into a pocket. A nylon strap is attached for easy handling. The lamps are available in threeMais informação do produto