Print…
silv-Air 7312 Hygiene Masks
Fornecedor: UVEX
8707.312
UVEX8707.312EA 33.8 EUR
UVEX8707.312
silv-Air 7312 Hygiene Masks
Máscaras Máscaras Higiénicas
This FFP3 preformed mask with an exhalation valve, featuring an innovative design and an optimised shape.
  • Four-point headband attachment for a comfortable, individual and secure fit
  • Extra-wide fabric headband
  • Ultrafine porous sealing lip
  • Nose clip integrated into the material for comfortable and secure positioning of the mask
The three- dimensional design enables a perfect fit for the many different face shapes which exist.