Print…

Anti-NUCL Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 130CT43.8.5]

Fornecedor: US Biological
031168-PE
USBI031168-PEEA 884 EUR
USBI031168-PE
Anti-NUCL Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 130CT43.8.5]
Anticorpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:500-1000; Immunochemistry: 1:50-100; Immunofluorescence: 1:10-50
Do not freeze! Light sensitive

Type: Primary
Antigen: NUCL
Clonality: Monoclonal
Clone: 130CT43.8.5
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR