Print…
Micro-splitter valves, Upchurch Scientific®
Fornecedor: UPCHURCH SCIENTIFIC
P-450
UPCHP-450EA 219 EUR
UPCHP-450
Micro-splitter valves, Upchurch Scientific®
Válvulas Micro-Válvulas
Designed to accurately split and control the low flow stream of a single incoming supply.
  • For interfacing LC-MS systems
  • Pressure rated to 800 psi (55 bar)
  • Adjustable split stream flow rates