Print…
Hot wire kit, Testo 440
Fornecedor: Testo
0563.4400
TEST0563.4400EA 552 EUR
TEST0563.4400
Hot wire kit, Testo 440
Registadores de dados
Hot wire kit equipped with clearly structured measurement menu for volume flow, parallel determination of air velocity, and air temperature in ventilation ducts.
  • Absolute pressure compensation, internal memory for data storage, and USB port for data export
  • Versstile and convenient integrated telescope with scaling
  • Easy to carry out all kinds of measurements
Ideal for long-term monitoring of indoor air quality and maximum flexibility for duct and outlet measurements.
Fornecimento: Supplied with testo 440 Hot Wire Kit, air velocity and IAQ measuringMais informação do produto