Print…
Cole-Parmer® FSB-200 Series Compact Fluidised Sand Baths
Fornecedor: Cole-Parmer
01189-50 01184-00 01184-02
TECN01189-50EA 9050 EUR
TECN01189-50 TECN01184-00 TECN01184-02
Cole-Parmer® FSB-200 Series Compact Fluidised Sand Baths
Banhos Banhos fluidizados
Fast heat-ups and accurate temperature control to 600 °C.
  • Dry, inert, non-toxic aluminium oxide ensures hazard-free operation
  • Safer alternative to high temperature oil and salt baths
  • Requires controller in order to be calibrated
  • Material: Stainless steel
Use these fluidised sand baths for heat treating processes, temperature sensor calibrations and electrical component testing. Baths use aluminium oxide fluidised by low-pressure air as a dry bath medium with excellent heat transfer properties. Inert, non-flammable aluminium oxide is safer to use thanMais informação do produto