Print…
Mini filters, TCI OPTI-SOLV™
Fornecedor: TCI
S1455-3CLM
TCIAS1455-3CLMEA 144 EUR
TCIAS1455-3CLM
Mini filters, TCI OPTI-SOLV™
Filtros Filtros de Pré-Coluna
The TCI OPTI-SOLV™ mini filters are similar in shape to TCI OPTI-GUARD™ Fits, and these filters not have any packing materials.
  • Provide low-impact filtration