Print…
96 Well Plate, RCH CEREX PWAX
Fornecedor: Tecan
694-0201RCH
TCAT694-0201RCHEA 0 EUR
TCAT694-0201RCH
96 Well Plate, RCH CEREX PWAX
Microplacas Placas de Extração em Fase Sólida
A 18 µm polymer micro particulate with weak anionic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep.
  • Cerex
  • Ion-exchange
  • Liquid handler compatible
Embalagem: 1 piece per box