Print…
SPE Plates, 96-well, NBE WWP2
Fornecedor: Tecan
417-0051NBE
TCAT417-0051NBEEA 0 EUR
TCAT417-0051NBE
SPE Plates, 96-well, NBE WWP2
Microplacas Placas de Extração em Fase Sólida
Polymeric water-wettable resin ideal for reverse-phase retention of aqueous samples.
  • Reversed-phase
  • Liquid handler compatible
Special water wettable polymer with 300 Å pore size ideal for protein digest clean up and other protein related workflows.