Print…
Single tissue blots, multiple species
Fornecedor: G Biosciences
TB28 TB29 TB30 TB31 TB32 TB33 TB34 TB35 TB36
TB28EA 1070 EUR
TB28 TB29 TB30 TB31 TB32 TB33 TB34 TB35 TB36
Single tissue blots, multiple species
Electroforese, Reagentes Western Blotting Reagents
Single tissue and multiple species blot contains four protein lanes with an identical GenLysate™ tissue lysate from human, mouse and rat species, and a protein marker lane.
  • Ready to screen
  • Single tissue multiple species (human, mouse, rat) blot
The premade blots are prepared with the GenLysate™ and then 50μg protein is loaded onto a 4 to 20% denaturing polyacrylamide gel, along with a prestained molecular weight marker. After the proteins are resolved, they are transferred to a PVDF membrane using Western Transfer Buffer. The blots are ready to be blocked and probed with the antibodies of choice.
Encomendar
ESPECIFICAÇÕES
Environmentally Preferable