Print…
Cole Parmer® Stuart MP-800 Automatic Digital Melting Point Apparatus
Fornecedor: Cole Parmer
03013-02 03013-03
STUA03013-02EA 8100 EUR
STUA03013-02 STUA03013-03
Cole Parmer® Stuart MP-800 Automatic Digital Melting Point Apparatus
Aparelhos para determinação do ponto de fusão
Capture and display video image of the melt on touch-screen display or download to a PC.
  • Accurately identify the melting point with advanced digital image processing
  • Get up and running quickly-intuitive setup and navigation on large 7" colour touch screen
  • Increase productivity-analyse three samples simultaneously
  • 8 GB storage holds approximately 300 melt files including videos
  • Set ramp rate from 0,1 to 20 °C in 0,1 °C increments
Advanced features with a familiar interface make this the ideal melting point apparatus for pharmaceutical, raw material andMais informação do produto