Print…
EasySep™ Mouse Pan-DC Enrichment Kit II
Fornecedor: STEMCELL TECHNOLOGIES
19863 19863RF
STMC19863EA 1700 EUR
STMC19863 STMC19863RF
EasySep™ Mouse Pan-DC Enrichment Kit II
Meios de separação celular
Immunomagnetic negative selection kit
  • Fast, easy-to-use, and column-free
  • Up to 65% purity
  • Isolated cells are untouched
  • Optimized for high yield
  • Facilitates rapid flow sorting of DC subsets
The EasySep™ Mouse Pan-DC Enrichment Kit II is designed to isolate dendritic cells (DCs) from mouse splenocytes by immunomagnetic negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labelled cells are separated from the untouched desired cells using an EasySep™ magnet and simplyMais informação do produto