Print…
EasySep™ Human Pan-Granulocyte Isolation Kits
Fornecedor: STEMCELL TECHNOLOGIES
19259 19259RF
STMC19259EA 2070 EUR
STMC19259 STMC19259RF
EasySep™ Human Pan-Granulocyte Isolation Kits
Meios de separação celular
Immunomagnetic isolation of untouched human pan-granulocytes
  • Fast, easy-to-use and column-free
  • Up to 99% purity
  • Untouched, viable cells
The EasySep™ Human Pan-Granulocyte Enrichment Kit is designed to isolate granulocytes (eosinophils, basophils and neutrophils) from normal peripheral whole blood by negative selection. Undesired cells are targeted for removal with Tetrameric Antibody Complexes recognising non-granulocytes and magnetic particles. The labelled cells are separated using an EasySep™ magnet without the use of columns. Desired cells areMais informação do produto