Print…
EasySep™ EasyStand™ Linkable Magnet Stands
Fornecedor: STEMCELL TECHNOLOGIES
18134 18136 18130
STMC18134EA 2810 EUR
STMC18134 STMC18136 STMC18130
EasySep™ EasyStand™ Linkable Magnet Stands
Ímanes
Linkable stands for EasySep™ magnets available in one, four, or six piece configurations.
  • Simplify the separation of one or more samples at one time
  • Magnets not included
Each EasySep™ EasyStand™ can hold a single EasySep™ magnet, and a simple system links up to 6 individual EasySep™ EasyStands™ together. After each round of magnetic separation, the supernatants from all samples are poured off simultaneously into the receiving tubes.
Atenção: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form,Mais informação do produto