Print…
EasySep™ Multistand
Fornecedor: STEMCELL TECHNOLOGIES
18010
STMC18010EA 2350 EUR
STMC18010
EasySep™ Multistand
Ímanes
The EasySep™ Multistand allows the separation of up to four samples at one time.
  • Immunomagnetic column-free magnet
An ingenious reversible clamping bar allows the multistand to hold up to 4 EasySep™ magnets or 4 EasySep™ 'The Big Easy' magnets. After each round of magnetic separation, the supernatants from all samples are poured off simultaneously into receiving tubes. The receiving tube bar is indexed to ensure proper position of the tubes.
Informações de encomenda: Magnets must be purchased separately.
Atenção: Product restrictions: (i)Mais informação do produto
Encomendar
ESPECIFICAÇÕES
Environmentally Preferable