Print…
RosetteSep™ Human CD4 Depletion Cocktails
Fornecedor: STEMCELL TECHNOLOGIES
15622 15662
STMC15622EA 238 EUR
STMC15622 STMC15662
RosetteSep™ Human CD4 Depletion Cocktails
Meios de separação celular
Immunodensity depletion of CD4+ cells.
  • Fast and easy-to-use
  • Requires no special equipment or training
  • Untouched, viable cells
  • Can be combined with SepMate™ for consistent, high-throughput sample processing
The RosetteSep™ Human CD4 Depletion Cocktail is designed to deplete CD4+ cells from whole blood. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognising CD4 and glycophorin A on red blood cells (RBCs). When centrifuged over a buoyant density medium such as Lymphoprep™ (STMC07801), the unwanted cells pellet along withMais informação do produto