Print…
EasySep™ Complete Kits for Human Whole Blood CD34+ Cells
Fornecedor: STEMCELL TECHNOLOGIES
15086RF 15086
STMC15086RFEA 1800 EUR
STMC15086RF STMC15086
EasySep™ Complete Kits for Human Whole Blood CD34+ Cells
Meios de separação celular
Immunomagnetic positive selection kit.
  • Fast and easy-to-use
  • Up to 98% purity
  • No columns required
  • Can be combined with SepMate™ for consistent, high-throughput sample processing
The Complete Kit for Human Whole Blood CD34+ Cells is a simple two-step method that is designed to isolate CD34+ cells from whole blood or buffy coat.
First, hematopoietic progenitor cells are pre-enriched using the RosetteSep™ Human Hematopoietic Progenitor Cell Enrichment Cocktail (15186C) with antibodies recognising CD2, CD3, CD14, CD16, CD19, CD24, CD56, CD66b and CD61Mais informação do produto