Print…
EasySep™ Human NK Cell Isolation Kits
Fornecedor: STEMCELL TECHNOLOGIES
17955RF 17955 100-0960
STMC17955RFEA 2030 EUR
STMC17955RF STMC17955 STMC100-0960
EasySep™ Human NK Cell Isolation Kits
Meios de separação celular
Immunomagnetic isolation of untouched human NK cells
  • Fast, easy-to-use and column-free
  • Up to 94% purity with high recovery
  • Isolated cells are untouched
The EasySep™ Human NK Cell Isolation Kit is designed to isolate NK cells from fresh or previously frozen peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labelled cells are separated from the untouched desired cells by using anMais informação do produto