Print…
Molecular model: silicon dioxide, molymod®
Fornecedor: SPIRING ENTERPRISES
MKO-136-38
SPIRMKO-136-38EA 0 EUR
SPIRMKO-136-38
Molecular model: silicon dioxide, molymod®
Modelos Biologia molecular
Molecular set with 38 atoms and connections for self-installation.
Molymod is a unique dual-scale system of high quality molecular models.