Print…
Resin Column Water Purifiers
Fornecedor: SMEG
WP3010
SMEGWP3010EA 3880 EUR
SMEGWP3010
Resin Column Water Purifiers
Sistemas para purificação da água Desionizadores
The water purifier can be used to produce deionised water at 0,8 to 1 μS/cm, suitable for the final rinsing of instrument washers. The system uses mixed bed resins (disposable) which can also retain silica. When the resins run out, they can simply be replaced with the MI700WP kit.