Print…
Knee Protector, Standard
Fornecedor: SIOEN
YMB00000C0
SIOEYMB00000C0EA 5.2 EUR
SIOEYMB00000C0
Knee Protector, Standard
Suportes ergonómicos Joelheiras
These knee protectors are recommended for work in the kneeling position. The protection distributes forces evenly and prevents small hard objects on the ground causing injuries.
Without knee protectors, workers may suffer discomfort and immediate injuries from hard surfaces and small stones and similar objects lying on the surfaces.
Certificações: Tested according to EN 14.404+A1:2010