Print…
Estojos porta-lâminas, LockMailer™
Fornecedor: Simport Scientific
M950-4MA
SIMPM950-4MAEA 414 EUR
SIMPM950-4MA
Estojos porta-lâminas, LockMailer™
Lâminas, Sistemas para envio
Transparent container made of high density polyethylene for mailing or storing slides between the doctor's office and the lab.
  • Inside channels slotted to keep slides separated
  • Unique tamper evident leakproof screw cap
  • Good stability on a bench