Print…
Viscosity flow cups
Fornecedor: Industrial Physics
VF2196 VF2197 VF2198 VF2195
SIMEVF2196EA 180.79 EUR
SIMEVF2196 SIMEVF2197 SIMEVF2198 SIMEVF2195
Viscosity flow cups
Viscosímetros Copos de fluxo
These viscosity flow cups are titanium anodised aluminum viscosity cups.
Suitable to be used with stand, to measure the viscosity of lacquers, paint and other liquids.
Certificações: AFNOR
Encomendar
ESPECIFICAÇÕES
Environmentally Preferable