Print…
Multi-finger scratch tester, Taber® TB2100
Fornecedor: Industrial Physics
TB2100
SIMETB2100EA 0 EUR
SIMETB2100
Multi-finger scratch tester, Taber® TB2100
Medidores de resistência
The Taber® multi-finger scratch / mar tester is used to measure the relative resistance or susceptibility of a material surface to scratching, marring, gouging, scraping, engraving and other similar physical damage that can’t be classified as ordinary wear.
  • Pneumatically driven, moveable platform
  • Control knob operation
  • Five independent spline-shaft fingers with support rest
  • 9-piece weight set (2N, 3N, 4.5N, 5N, 6N, 7N, 10N, 15N, 20N)
  • "Gripping" spring clamp specimen hold-down
  • Replaceable specimen platform protective guard
  • Electronic timer accurate to 1/100