Print…
Reference guide and photographs for steel surfaces, SSPC-VIS 4/NACE VIS 7
Fornecedor: Industrial Physics
LD3058
SIMELD3058EA 459 EUR
SIMELD3058
Reference guide and photographs for steel surfaces, SSPC-VIS 4/NACE VIS 7
Livros
The SSPC-VIS 4/NACE VIS 7 shows six different steel surfaces - two uncoated and four previously coated - before and after waterjetting.
  • Photographs illustrate four separate degrees of cleaning (WJ 1, WJ 2, WJ 3, and WJ 4) for each initial condition
  • Additional photos that depict the appearance light, moderate, and heavy flash rust after cleaning
  • Cleanliness requirements for conditions depicted are defined
  • Joint SSPC/NACE surface preparation specification for high and ultra high pressure water jetting (SP 12/NACE 5)
Informações de encomenda: Contains aMais informação do produto