Print…
Scientific Calculator
Fornecedor: Merck
C3923-1EA C8474-1EA
SIALC3923-1EAEA 114 EUR
SIALC3923-1EA SIALC8474-1EA
Scientific Calculator
Calculadoras Calculadoras de bolso
Solar powered calculator with protective sliding case.
Functions: Mathematics - Exponential - Trigonometry - Inverse trigonometry - Statistical functions (mean and standard deviation); Angle/time conversions.