Print…

(±)-2,3-Epoxypropyl methacrylate

Fornecedor: Merck
Atenção

Sinônimos: Oxiran-2-ylmethyl methacrylate, Glycidyl methacrylate

779342-100ML
SIAL779342-100MLEA 31 EUR
SIAL779342-100ML
(±)-2,3-Epoxypropyl methacrylate
(±)-2,3-Epoxypropyl methacrylate
Fórmula: C₇H₁₀O₃
Peso Molecular: 142.15 g/mol
Número MDL : MFCD00005137
Número CAS: 106-91-2
EINECS: 203-441-9
UN: 2810
ADR: 6.1 III

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR