Print…
HyClone™ Newborn Bovine Calf Serum, US Origin, Cytiva
Fornecedor: HyClone products (Cytiva)
SH30118.03 SH30118.02 SH30118.04 SH30118.02HI SH30118.03HI SH30118.04HI SH30118.03IR25
HYCLSH30118.03EA 121 EUR
HYCLSH30118.03 HYCLSH30118.02 HYCLSH30118.04 HYCLSH30118.02HI HYCLSH30118.03HI SH30118.04HI SH30118.03IR25
HyClone™ Newborn Bovine Calf Serum, US Origin, Cytiva
Soro
Bovine calf serum products are excellent and cost-effective alternatives to fetal bovine serum (FBS). Bovine calf serum products contain exceptionally high levels of transferrin, which can provide three to four times as much available iron as FBS.
  • Complete traceability back to original source
  • Low in antibodies and high in growth factors
  • Virus panel testing according to 9 CFR 113.53
HyClone™ Newborn Bovine Calf Serum, US Origin undergoes the same careful collection and processing procedures (including venipuncture) used for our bovine serum products.Mais informação do produto