Print…
Funnel with ball and lid, ARNOLD, V2.0
Fornecedor: S.C.A.T.
317642 317641 317632 317643 317644 317645 317646 317647 317648 317649 319001 317636 317660
221-0298EA 181 EUR
221-0298 SCTV317641 SCTV317632 SCTV317643 SCTV317644 SCTV317645 SCTV317646 SCTV317647 SCTV317648 SCTV317649 SCTV319001 SCTV317636 SCTV317660
Funnel with ball and lid, ARNOLD, V2.0
Funis Funis de segurança
The strong safety funnel 'ARNOLD' V2.0 with lid and ball meets the highest requirements for safe disposal of hazardous liquid waste due to its combination of two independent safety mechanisms. The two safety features consist of a ball integrated in the funnel basin and a sealing ring incorporated in the hinged lid of the funnel, which prevent the escape of toxic or explosive gases. The new design ensures greater stability, durability and prevents deformation of the funnel.
  • Made of high quality white/blue HDPE material for maximum stability and durability