Print…
Continuous level control, SymLine®
Fornecedor: S.C.A.T.
199005 199013 160123 502042 108042-S1 108178
SCTV199005EA 35 EUR
SCTV199005 216-0923 SCTV160123 SCTV502042 SCTV108042-S1 SCTV108178
Continuous level control, SymLine®
Sensores de nível
Measuring filling levels continuously. Monitoring non-transparent canisters on a permanent basis is no problem with the continuous level control unit.
  • Automatic warning for 5 different filling levels freely adjustable, with accuracy of up to ±0,5 mm
  • Control box provides 5 freely configurable outlets to control external devices
  • Disc sensor for level control from non-conductive material is easy to install and adjustable in height
  • Rod sensor and canister with removable sleeve allow easy maintenance and cleaning
  • Canister made from electrostatic conductive HDPE
Mais informação do produto