Print…
Cannon-Fenske viscometers for automatic measurements
Fornecedor: SI Analytics
285403712 285403729 285403745 285403753 285403761 285403778 285403786 285403794 285403807 285403737 285403815
SCOI285403712EA 381 EUR
SCOI285403712 SCOI285403729 SCOI285403745 SCOI285403753 SCOI285403761 SCOI285403778 SCOI285403786 SCOI285403794 SCOI285403807 SCOI285403737 SCOI285403815
Cannon-Fenske viscometers for automatic measurements
Viscosímetros Viscosímetros capilares
Cannon-Fenske viscometers for automatic measurements especially in routine analyses.
Suitable for transparent liquids, foaming liquids, and liquid mixture with highly volatile components.