Print…
Electrode Plug Cable Combinations
Fornecedor: SI Analytics
285124613 285124654 285124708
SCOI285124613EA 53.7 EUR
SCOI285124613 SCOI285124654 SCOI285124708
Electrode Plug Cable Combinations
Cabos
Electrode cables along with electrode socket, 1 m two core cable up to 80 °C, for resistance thermometer and 2-pole-conductivity cell without temperature sensor. Available with different cable length.