Print…
Fibre optic ring lights
Fornecedor: SCHOTT AG Lighting and imaging
157066 157081 157086
SCOC157066EA 731 EUR
SCOC157066 SCOC157081 SCOC157086
Fibre optic ring lights
Luzes Ring Lights
Fibre optic ring lights are available as six point, annular, or darkfield versions with specific advantages. For all ring lights, adapters are available for a variety of objective series. Ring lights are used to provide intense, uniform, and shadow-free illumination.
  • Mounts directly to the objective using a thumb screw
  • Housing made of rugged, black, anodised aluminium
  • Fibre bundle protected by flexible metal PVC sheathing
  • High white light transmission thanks to the use of SCHOTT PURAVIS ® fibres
Annular ring lights offer absolute shadow-freeMais informação do produto