Print…
Donor horse serum
Fornecedor: Biowest
S0900-100 S0900-500 S090H-100 S090H-500 S0900-1000 S090Y-100 S090Y-500 S090G-100
S0900-100EA 20.8 EUR
S0900-100 S0900-500 S090H-100 S090H-500 BWSTS0900-1000 S090Y-100 S090Y-500 S090G-100
Donor horse serum
Soro
Donor horse serum is aseptically collected from herds of healthy animals inspected periodically by a licensed veterinarian. Applications for donor horse serum include mycoplasma contamination tests carried out on media, blocking agent and controls in immunoassays, culture of PC12 cells when combined with FBS.