Print…
Fetal bovine serum, heat inactivated, certified, U.S. origin
Fornecedor: Rockland Immunochemicals
FBS-01-0100
ROCKFBS-01-0100EA 341 EUR
ROCKFBS-01-0100
Fetal bovine serum, heat inactivated, certified, U.S. origin
Soro
Fatal bovine serum is used in a wide range of cell culture applications.
  • Sterile bacteriophage
  • Store at −20 °C
This bovine serum is certified to originate in the United States as heat inactivated, performance, mycoplasma, virus, bacteriophage and endotoxin tested.
Atenção: For in vitro diagnostic use only, not for human or animal therapeutic use.
Encomendar
ESPECIFICAÇÕES
Environmentally Preferable
Endotoxinas <1,0 EU/ml