Print…

Anti-IgG (H+L) Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Fornecedor: Rockland Immunochemicals
707-402-002
ROCK707-402-002EA 341 EUR
ROCK707-402-002
Anti-IgG (H+L) Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorpos
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring extremely low background levels, absence of F(c) mediated binding, lot-to-lot consistency, high titer and specificity.

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 500 µl

Type: Secondary
Antigen: IgG (Golden Syrian Hamster) (Golden Syrian Hamster IgG)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)Epitope:
Host: Rabbit
Isotype: none
Reactivity: Hamster
Encomendar

ESPECIFICAÇÕES

Símbolo do antígeno IgG (H+L)
Environmentally Preferable
Tipo de anticorpo Secundário
Clonalidade Policlonal
Conjugado FITC (Fluorescein Isothiocyanate)
Reatividade Golden syrian hamster
Hospedeiro Rabbit
Isótipo
Western blot Yes
ELISA Yes
ImunoQuímica Yes
ImunoPrecipitação Yes
ImunoFluorescência Yes
Mudança de gel Yes
Citometria de fluxo Yes
ChIP Yes
Neutralisation Yes
Fragmento de anticorpo F(ab')2
Imunogênio Golden Syrian Hamster IgG whole molecule
Formato Antibody fraction
Forma Lyophilized
Purification purified by immunoaffinity chromatography
Concentração 1,0 mg/ml by UV absorbance at 280 nm
Adsorção cruzada none
Storage buffer 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free
Temperatura de armazenamento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR