Print…

Anti-BDNF Rabbit Polyclonal Antibody

Fornecedor: Rockland Immunochemicals
209-401-C27
ROCK209-401-C27EA 821 EUR
ROCK209-401-C27
Anti-BDNF Rabbit Polyclonal Antibody
Anticorpos
This purified antibody has been tested for use in ELISA and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 27 kDa in size corresponding to the mature human BDNF Protein by western blotting.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:1000;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: BDNF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR