Print…
Cover mat for deep 96-well plates
Fornecedor: RATIOLAB
8101101
RATI8101101EA 360 EUR
RATI8101101
Cover mat for deep 96-well plates
Vedantes Selos de microplaca
Closures for 96-Well Deep-Well-Plates Low-Profile, 1,2 ml. Made of thermo-plastic elastomer (TPE).
Autoclavable at 121 °C, 20 minutes.