Print…
Autoclavable Bags, Biohazard
Fornecedor: RATIOLAB
7003220
RATI7003220EA 461 EUR
RATI7003220
Autoclavable Bags, Biohazard
Sacos Sacos de Eliminação de Resíduos
Autoclavable PP bags for the disposal of contaminated labware.
  • Distinctive red colour for easy identification
  • Rectangle white indicator patch field turns into dark when autoclaved
  • BIOHAZARD symbol and safety imprint in German, English and French language
  • Autoclavable at 134 °C (20 minutes)
Atenção: The bag must not be completely closed when autoclaving.